Gene Transcripts
- Gene Symbol
- Cnpy3
- Entrez Gene ID
- 72029
- Gene Name
- canopy 3 homolog (zebrafish)
- Gene Aliases
- 1600025D17Rik, 2410050O22Rik, AI413153, CAG4A, ERDA5, PRAT4A, Tnrc5
- Chromosome Location
- Chr. 17: 46735713 - 46752293 on Build GRCm38
- UniGene ID
- Mm.240325
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF361644.1 |
- |
3, 4 |
464 |
|
AK005532.1 |
- |
3, 4 |
447 |
|
AK031742.1 |
- |
3, 4 |
302 |
|
AK049358.1 |
- |
3, 4 |
450 |
|
AK082749.1 |
- |
1, 2 |
66 |
|
AK085617.1 |
- |
3, 4 |
457 |
|
AK086940.1 |
- |
3, 4 |
397 |
|
AK143428.1 |
- |
3, 4 |
440 |
|
AK220209.1 |
- |
3, 4 |
274 |
|
BB857917.1 |
- |
3, 4 |
467 |
|
BC013549.1 |
- |
3, 4 |
466 |
4222133 |
View Full Product Details