Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027045.1 | NP_081321.1 | 11 | 2933 | |
NM_028407.3 | NP_082683.2 | 8 | 1020 | ||
GenBank mRNA | AK005931.1 | - | 8 | 1020 | |
AK147383.1 | - | 11 | 2928 | ||
BC111465.1 | - | 9 | 1229 |