Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028773.3 | NP_083049.1 | 7 | 1072 | |
GenBank mRNA | AJ306422.1 | - | 7 | 938 | |
AK004734.2 | - | 7 | 1073 | ||
AK156202.1 | - | 7 | 1137 | ||
AK171017.1 | - | 7 | 1070 | ||
BC028773.1 | - | 4 | 631 | ||
BC064734.1 | - | 7 | 1057 |