Gene Transcripts
- Gene Symbol
- Pck2
- Entrez Gene ID
- 74551
- Gene Name
- phosphoenolpyruvate carboxykinase 2 (mitochondrial)
- Gene Aliases
- 1810010O14Rik, 9130022B02Rik, PEPCK-M
- Chromosome Location
- Chr. 14: 55540263 - 55550777 on Build GRCm38
- UniGene ID
- Mm.29856
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028994.2 |
NP_083270.1
|
9, 10 |
1685 |
|
GenBank mRNA |
AK007429.1 |
- |
6, 7 |
877 |
|
AK030501.1 |
- |
9, 10 |
1686 |
|
AK031612.1 |
- |
9, 10 |
1530 |
|
AK034927.1 |
- |
9, 10 |
1564 |
|
AK142561.1 |
- |
3, 4 |
437 |
|
AK147972.1 |
- |
9, 10 |
1539 |
|
AK165094.1 |
- |
9, 10 |
1563 |
|
AK170242.1 |
- |
9, 10 |
1539 |
|
AK218547.1 |
- |
2, 3 |
224 |
|
BC010318.1 |
- |
9, 10 |
1548 |
3596227 |
BC011126.1 |
- |
8, 9 |
1847 |
|
BC023437.1 |
- |
7, 8 |
1153 |
|
BC028326.1 |
- |
4, 5 |
486 |
|
View Full Product Details