Gene Transcripts

Gene Symbol
Ankrd7
Entrez Gene ID
75196
Gene Name
ankyrin repeat domain 7
Gene Aliases
4930532L20Rik, AV278427
Chromosome Location
Chr. 6: 18861634 - 18879586 on Build GRCm38
UniGene ID
Mm.67665
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167757.1 NP_001161229.1 3 571
NM_029202.2 NP_083478.2 3 568
GenBank mRNA AK015948.1 - 3 512
AK077080.1 - 3 540
BC049731.1 - 3 568 6771078

View Full Product Details