Gene Transcripts

Gene Symbol
Ndel1
Entrez Gene ID
83431
Gene Name
nuclear distribution gene E-like homolog 1 (A. nidulans)
Gene Aliases
2600006O07Rik, MITAP1, NUDEL, mNudel
Chromosome Location
Chr. 11: 68821434 - 68853131 on Build GRCm38
UniGene ID
Mm.31979
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023668.2 NP_076157.2 5 654
GenBank mRNA AF290472.1 - 5 586
AF323918.1 - 5 674
AK011168.1 - 5 564
AK146029.1 - 5 583
BC021434.1 - 550 5029832
BC046796.1 - 5 585 5718032

View Full Product Details