Gene Transcripts

Gene Symbol
Ccdc86
Entrez Gene ID
108673
Gene Name
coiled-coil domain containing 86
Gene Aliases
4933411H20Rik, 6720480F16Rik, D19Ertd678e
Chromosome Location
Chr. 19: 10941481 - 10949266 on Build GRCm38
UniGene ID
Mm.294908
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023731.3 NP_076220.2 3, 4 1182
GenBank mRNA AB041663.1 - 3, 4 1151
AK016771.1 - 3, 4 558
AK036812.1 - 3, 4 1173
AK078407.1 - 3, 4 1183
AK078455.1 - 3, 4 1183
AK088727.1 - 3, 4 1175
BC043326.1 - 3, 4 1178 5374087
DQ501252.1 - 3, 4 1149

View Full Product Details