Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166368.1 | NP_001159840.1 | 2 | 181 | |
NM_008020.3 | NP_032046.1 | 2 | 196 | ||
GenBank mRNA | AK144284.1 | - | 2 | 197 | |
AK165394.1 | - | 2 | 161 | ||
BC031824.1 | - | 2 | 150 | 4196363 | |
BC053692.1 | - | 2 | 186 | 6395841 | |
DV655383.1 | - | 2 | 181 |