Gene Transcripts

Gene Symbol
Klra2
Entrez Gene ID
16633
Gene Name
killer cell lectin-like receptor, subfamily A, member 2
Gene Aliases
Klra30, Ly49b
Chromosome Location
Chr. 6: 131198976 - 131247723 on Build GRCm38
UniGene ID
Mm.4783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170851.1 NP_001164322.1 6 812
NM_008462.5 NP_032488.4 6 812
GenBank mRNA AF253057.1 - 5 646
AF253058.1 - 5 646
AF253059.1 - 5 646
AF395446.1 - 5 646
AK017140.1 - 6 803
AY860971.1 - 5 646
BC064711.1 - 7 812 30295755
EF025059.1 - 6 646
U10304.1 - 6 740

View Full Product Details