Gene Transcripts

Gene Symbol
Zfp87
Entrez Gene ID
170763
Gene Name
zinc finger protein 87
Gene Aliases
2210039O17Rik, 4631412O18Rik, KRAB4, Mzf22, Zfp72
Chromosome Location
Chr. 13: 67515781 - 67526231 on Build GRCm38
UniGene ID
Mm.491205
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133228.3 NP_573491.2 4 1517
GenBank mRNA AF242377.1 - 4 1420
AK028465.1 - 1 1073
AK030724.1 - 4 1465
AK049711.1 - 4 1437
BC031988.1 - 4 1420 4017149

View Full Product Details