Gene Transcripts

Gene Symbol
Ppp2ca
Entrez Gene ID
19052
Gene Name
protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform
Gene Aliases
PP2A, R75353
Chromosome Location
Chr. 11: 52098824 - 52122749 on Build GRCm38
UniGene ID
Mm.260288
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019411.4 NP_062284.1 2 433
GenBank mRNA AK076110.1 - 2 434
AK172644.1 - 2 406
BC003856.1 - 2 410 3484084
BC054458.1 - 2 393 6493866
Z67745.1 - 2 208

View Full Product Details