Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163307.1 | NP_001156779.1 | 8 | 735 | |
NM_001163308.1 | NP_001156780.1 | 7 | 597 | ||
NM_009545.2 | NP_033571.1 | 10 | 1002 | ||
GenBank mRNA | AF483502.1 | - | 6 | 443 | |
AF483503.1 | - | 6 | 443 | ||
AK142837.1 | - | 7 | 510 | ||
BC016419.1 | - | 8 | 689 | 4458285 | |
D90085.1 | - | 10 | 850 |