Gene Transcripts

Gene Symbol
Lrp11
Entrez Gene ID
237253
Gene Name
low density lipoprotein receptor-related protein 11
Gene Aliases
1700034J19Rik, 6330533B21, 9830160H19Rik
Chromosome Location
Chr. 10: 7589767 - 7625483 on Build GRCm38
UniGene ID
Mm.206759
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172784.3 NP_766372.1 7 1629
GenBank mRNA AK031994.1 - 7 1571
AK036676.1 - 7 1612
AK050544.1 - 7 1599
BC059874.1 - 7 1446

View Full Product Details