Gene Transcripts

Gene Symbol
Lrrc30
Entrez Gene ID
240131
Gene Name
leucine rich repeat containing 30
Gene Aliases
Gm326
Chromosome Location
Chr. 17: 67630965 - 67632723 on Build GRCm38
UniGene ID
Mm.295736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033340.3 NP_001028512.2 1 503
GenBank mRNA AK137598.1 - 1 504
BC147419.1 - 1 446 9056500
BC147420.1 - 1 446 9056487

View Full Product Details