Gene Transcripts

Gene Symbol
Cops7b
Entrez Gene ID
26895
Gene Name
COP9 (constitutive photomorphogenic) homolog, subunit 7b (Arabidopsis thaliana)
Gene Aliases
D1Wsu66e, E130114M23
Chromosome Location
Chr. 1: 86582902 - 86606500 on Build GRCm38
UniGene ID
Mm.12973
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172974.2 NP_766562.1 2 261
GenBank mRNA AF071317.1 - 2 239
AK053615.1 - 2 220
AK081309.1 - 2 261
AK168740.1 - 2 220
AK170140.1 - 2 220

View Full Product Details