Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025887.4 | NP_080163.1 | 2 | 556 | |
GenBank mRNA | AB232593.1 | - | 1 | 89 | |
AK002631.1 | - | 2 | 311 | ||
AK008198.1 | - | 2 | 297 | ||
AK010495.1 | - | 2 | 401 | ||
AK076093.1 | - | 2 | 537 | ||
AK081999.1 | - | 2 | 557 | ||
AK150296.1 | - | 2 | 310 | ||
AK150346.1 | - | 2 | 310 | ||
AK150405.1 | - | 2 | 354 | ||
AK151066.1 | - | 2 | 310 | ||
AK153131.1 | - | 2 | 310 | ||
AK159706.1 | - | 2 | 310 | ||
BC004842.1 | - | 2 | 417 | 2649317 | |
BC034370.1 | - | 2 | 299 | 3709729 | |
BC096481.1 | - | 3 | 490 | 5322132 |