Gene Transcripts

Gene Symbol
Vmn2r60
Entrez Gene ID
637898
Gene Name
vomeronasal 2, receptor 60
Gene Aliases
Casr-rs3, EG637898, Gprc2a-rs3
Chromosome Location
Chr. 7: 42116471 - 42195776 on Build GRCm38
UniGene ID
Mm.469900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105057.1 NP_001098527.1 2 238

View Full Product Details