Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178802.4 | NP_848917.2 | 6 | 1339 | |
GenBank mRNA | AK028857.1 | - | 6 | 1228 | |
AK047439.1 | - | 6 | 1339 | ||
AK050080.1 | - | 6 | 1251 | ||
AK079982.1 | - | 5 | 1421 | ||
BC118954.1 | - | 6 | 1094 | ||
BC118955.1 | - | 6 | 1094 |