Gene Transcripts

Gene Symbol
Plekhm1
Entrez Gene ID
353047
Gene Name
pleckstrin homology domain containing, family M (with RUN domain) member 1
Gene Aliases
AP162, B2, BC038943, D330036J23Rik
Chromosome Location
Chr. 11: 103365092 - 103412664 on Build GRCm38
UniGene ID
Mm.18556
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183034.1 NP_898855.1 4 543
GenBank mRNA AK154703.1 - 3 472
AK155357.1 - 4 567
AK157752.1 - 4 578
AK190944.1 - 1 67
BC053079.1 - 4 543 5702529

View Full Product Details