Gene Transcripts

Gene Symbol
Fam24a
Entrez Gene ID
68223
Gene Name
family with sequence similarity 24, member A
Gene Aliases
1700063I17Rik
Chromosome Location
Chr. 7: 131334622 - 131336716 on Build GRCm38
UniGene ID
Mm.233827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183272.2 NP_899095.1 3 168
GenBank mRNA AK006815.1 - 3 169
AK018853.1 - 3 170
AK018873.1 - 3 208
AK018966.1 - 3 208
BC048642.1 - 3 225 6772199

View Full Product Details