Gene Transcripts
- Gene Symbol
- Cwc22
- Entrez Gene ID
- 80744
- Gene Name
- CWC22 spliceosome-associated protein homolog (S. cerevisiae)
- Gene Aliases
- AA684037, AI173004, AL022752, B230213M24, mKIAA1604
- Chromosome Location
- Chr. 2: 77895653 - 77946356 on Build GRCm38
- UniGene ID
- Mm.288151
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290740.1 |
NP_001277669.1
|
6, 7 |
896 |
|
NM_030560.5 |
NP_085037.2
|
7, 8 |
954 |
|
NM_172667.2 |
NP_766255.1
|
6, 7 |
893 |
|
GenBank mRNA |
AK045589.1 |
- |
6, 7 |
725 |
|
AK077961.1 |
- |
7, 8 |
782 |
|
AK089988.1 |
- |
|
732 |
|
AK129404.1 |
- |
|
710 |
|
AK132001.1 |
- |
6, 7 |
837 |
|
AK143352.1 |
- |
6, 7 |
726 |
|
AK149528.1 |
- |
6, 7 |
720 |
|
AK166487.1 |
- |
6, 7 |
732 |
|
BC003993.1 |
- |
6, 7 |
706 |
3492146 |
View Full Product Details