Gene Transcripts

Gene Symbol
Fibcd1
Entrez Gene ID
98970
Gene Name
fibrinogen C domain containing 1
Gene Aliases
AI448887
Chromosome Location
Chr. 2: 31813290 - 31846647 on Build GRCm38
UniGene ID
Mm.32080
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178887.4 NP_849218.2 6 1217
GenBank mRNA AK046338.1 - 6 1179
AK084379.1 - 7 1220
BC060634.1 - 6 1217 6848779

View Full Product Details