Gene Transcripts

Gene Symbol
Cyp2c39
Entrez Gene ID
13098
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 39
Gene Aliases
CYPIIC39
Chromosome Location
Chr. 19: 39510822 - 39568529 on Build GRCm38
UniGene ID
Mm.42101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF047726.1 - 3 399
BC057911.1 - 3 409
BC139290.1 - 3 448 8862312
BC139291.1 - 3 448 8862313

View Full Product Details