Gene Transcripts

Gene Symbol
Fscn2
Entrez Gene ID
238021
Gene Name
fascin homolog 2, actin-bundling protein, retinal (Strongylocentrotus purpuratus)
Gene Aliases
A930022G03, Ahl8, C630046B20Rik
Chromosome Location
Chr. 11: 120359851 - 120368173 on Build GRCm38
UniGene ID
Mm.481403
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172802.4 NP_766390.2 1 739
GenBank mRNA AK044556.1 - 1 738
AK050002.1 - 1 740
AK078674.1 - 1 767
BC109356.1 - 1 700 40057445
BC109357.1 - 1 700 40057447

View Full Product Details