Gene Transcripts

Gene Symbol
Dock7
Entrez Gene ID
67299
Gene Name
dedicator of cytokinesis 7
Gene Aliases
3110056M06Rik, Gm430, m
Chromosome Location
Chr. 4: 98936659 - 99122605 on Build GRCm38
UniGene ID
Mm.260623
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290636.1 NP_001277565.1 16 2006
NM_026082.5 NP_080358.3 16 2006
GenBank mRNA BC094420.1 - 3 202
CA752561.1 - 3 204

View Full Product Details