Gene Transcripts

Gene Symbol
Btbd16
Entrez Gene ID
330660
Gene Name
BTB (POZ) domain containing 16
Gene Aliases
E330040A16Rik
Chromosome Location
Chr. 7: 130774069 - 130825899 on Build GRCm38
UniGene ID
Mm.271550
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081038.2 NP_001074507.2 15 1707
GenBank mRNA BY255452.1 - 4 462

View Full Product Details