Gene Transcripts

Gene Symbol
Ogfr
Entrez Gene ID
72075
Gene Name
opioid growth factor receptor
Gene Aliases
2010013E17Rik
Chromosome Location
Chr. 2: 180589407 - 180595837 on Build GRCm38
UniGene ID
Mm.250418
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031373.3 NP_113550.3 7 969
GenBank mRNA AF303894.1 - 7 917
AK008230.1 - 7 934
AK149146.1 - 7 916
AK150115.1 - 7 901
BC049965.1 - 7 915 3155091

View Full Product Details