Gene Transcripts

Gene Symbol
Klk9
Entrez Gene ID
101533
Gene Name
kallikrein related-peptidase 9
Gene Aliases
1200016C12Rik, AI324041
Chromosome Location
Chr. 7: 43791588 - 43796758 on Build GRCm38
UniGene ID
Mm.19942
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028660.3 NP_082936.2 4 655
GenBank mRNA BC109326.1 - 4 567
BC109327.1 - 4 567 40051200

View Full Product Details