Gene Transcripts
- Gene Symbol
- Fam73b
- Entrez Gene ID
- 108958
- Gene Name
- family with sequence similarity 73, member B
- Gene Aliases
- 5730472N09Rik, AA408683, AI790341, C9orf54, R74766
- Chromosome Location
- Chr. 2: 30364198 - 30385520 on Build GRCm38
- UniGene ID
- Mm.254974
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001242407.1 |
NP_001229336.1
|
4, 5 |
638 |
|
NM_175392.3 |
NP_780601.1
|
4, 5 |
806 |
|
GenBank mRNA |
AK077618.1 |
- |
4, 5 |
599 |
|
AK131152.1 |
- |
1, 2 |
1492 |
|
AK140122.1 |
- |
4, 5 |
624 |
|
AK154200.1 |
- |
4, 5 |
627 |
|
AK165996.1 |
- |
4, 5 |
629 |
|
AK210993.1 |
- |
2, 3 |
149 |
|
AK218176.1 |
- |
2, 3 |
169 |
|
BC051404.1 |
- |
4, 5 |
806 |
6395030 |
View Full Product Details