Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001165902.1 | NP_001159374.1 | 5 | 670 | |
NM_007614.3 | NP_031640.1 | 5 | 870 | ||
GenBank mRNA | AK018515.1 | - | 5 | 784 | |
AK020013.1 | - | 5 | 785 | ||
AK035311.1 | - | 5 | 783 | ||
AK132398.1 | - | 5 | 851 | ||
AK133656.1 | - | 5 | 783 | ||
AK135743.1 | - | 2 | 337 | ||
AY751549.1 | - | 5 | 795 | ||
BC043078.1 | - | 2 | 272 | ||
BC048153.1 | - | 5 | 795 | 5709247 | |
BC053065.1 | - | 5 | 795 | 5709247 | |
M90364.1 | - | 5 | 680 |