Gene Transcripts

Gene Symbol
Efna4
Entrez Gene ID
13639
Gene Name
ephrin A4
Gene Aliases
EFL-4, Epl4, LERK-4
Chromosome Location
Chr. 3: 89333393 - 89338028 on Build GRCm38
UniGene ID
Mm.16332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007910.2 NP_031936.2 2 222
GenBank mRNA AK012195.1 - 2 226
AK142604.1 - 2 223
U90663.1 - 2 222

View Full Product Details