Gene Transcripts

Gene Symbol
Gas1
Entrez Gene ID
14451
Gene Name
growth arrest specific 1
Gene Aliases
AW554192, Gas-1
Chromosome Location
Chr. 13: 60174405 - 60177535 on Build GRCm38
UniGene ID
Mm.22701
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008086.2 NP_032112.1 1 1035
GenBank mRNA BC055747.1 - 2 130
BC058628.1 - 2 675 6401957
X65128.1 - 1 865

View Full Product Details