Gene Transcripts

Gene Symbol
Gng8
Entrez Gene ID
14709
Gene Name
guanine nucleotide binding protein (G protein), gamma 8
Gene Aliases
G(y)8
Chromosome Location
Chr. 7: 16891737 - 16895451 on Build GRCm38
UniGene ID
Mm.103770
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010320.3 NP_034450.1 3 704
GenBank mRNA AK144609.1 - 3 705
BC111437.1 - 1 68 40049157
BQ174809.1 - 2 231

View Full Product Details