Gene Transcripts
- Gene Symbol
- Hp1bp3
- Entrez Gene ID
- 15441
- Gene Name
- heterochromatin protein 1, binding protein 3
- Gene Aliases
- Hp1bp74
- Chromosome Location
- Chr. 4: 138216582 - 138244683 on Build GRCm38
- UniGene ID
- Mm.348429
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001122896.2 |
NP_001116368.1
|
10, 11 |
1297 |
|
NM_001122897.2 |
NP_001116369.1
|
11, 12 |
1534 |
|
NM_001285478.1 |
NP_001272407.1
|
11, 12 |
1554 |
|
NM_001285479.1 |
NP_001272408.1
|
11, 12 |
1492 |
|
NM_001285480.1 |
NP_001272409.1
|
10, 11 |
1258 |
|
NM_001285481.1 |
NP_001272410.1
|
11, 12 |
1456 |
|
NM_010470.3 |
NP_034600.2
|
11, 12 |
1495 |
|
GenBank mRNA |
AK004938.1 |
- |
11, 12 |
1472 |
|
AK028181.1 |
- |
11, 12 |
1470 |
|
AK043260.1 |
- |
10, 11 |
1271 |
|
AK050234.1 |
- |
11, 12 |
1474 |
|
AK075725.1 |
- |
10, 11 |
1235 |
|
AK161041.1 |
- |
10, 11 |
1265 |
|
AK166161.1 |
- |
11, 12 |
1442 |
|
AK169740.1 |
- |
11, 12 |
1401 |
|
BC004053.1 |
- |
7, 8 |
775 |
|
BC020024.1 |
- |
11, 12 |
1445 |
3597151 |
View Full Product Details