Gene Transcripts

Gene Symbol
Inhbc
Entrez Gene ID
16325
Gene Name
inhibin beta-C
Gene Aliases
Chromosome Location
Chr. 10: 127356325 - 127370544 on Build GRCm38
UniGene ID
Mm.2594
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010565.3 NP_034695.1 2 598
GenBank mRNA AK131946.1 - 2 198
BC026140.1 - 2 496 4212175
U95962.1 - 2 598
X90819.1 - 2 581

View Full Product Details