Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010914.2 | NP_035044.1 | 3 | 279 | |
GenBank mRNA | AK010762.1 | - | 3 | 259 | |
AK011792.1 | - | 3 | 198 | ||
AK079209.1 | - | 3 | 234 | ||
AK146168.1 | - | 3 | 281 | ||
AK154749.1 | - | 3 | 241 | ||
AK155749.1 | - | 3 | 241 | ||
BC010719.1 | - | 3 | 220 | 2811674 | |
X55316.1 | - | 3 | 275 |