Gene Transcripts

Gene Symbol
Psmb1
Entrez Gene ID
19170
Gene Name
proteasome (prosome, macropain) subunit, beta type 1
Gene Aliases
AA409053, C81484, Lmpc5
Chromosome Location
Chr. 17: 15475721 - 15498276 on Build GRCm38
UniGene ID
Mm.32912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011185.3 NP_035315.1 1, 2 129
GenBank mRNA AK028265.1 - 1, 2 151
AK077520.1 - 1, 2 130
AK132161.1 - 1, 2 130
AK167647.1 - 1, 2 151
BC018351.1 - 1, 2 130 3584538
U60824.1 - 1, 2 144
X80686.1 - 1, 2 133

View Full Product Details