Gene Transcripts
- Gene Symbol
- Psme2
- Entrez Gene ID
- 19188
- Gene Name
- proteasome (prosome, macropain) activator subunit 2 (PA28 beta)
- Gene Aliases
- AA589371, AI788882, PA28b
- Chromosome Location
- Chr. 14: 55587440 - 55591146 on Build GRCm38
- UniGene ID
- Mm.371573
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001029855.1 |
NP_001025026.1
|
10, 11 |
669 |
|
NM_011190.3 |
NP_035320.1
|
10, 11 |
769 |
|
GenBank mRNA |
AA216967.1 |
- |
1, 2 |
48 |
|
AK012344.1 |
- |
10, 11 |
698 |
|
AK088654.1 |
- |
10, 11 |
696 |
|
AK143556.1 |
- |
10, 11 |
701 |
|
BC002301.1 |
- |
2, 3 |
112 |
|
BC005680.1 |
- |
10, 11 |
689 |
3154255 |
BC057859.1 |
- |
10, 11 |
766 |
5310526 |
BY498670.1 |
- |
4, 5 |
274 |
|
U60329.1 |
- |
10, 11 |
638 |
|
- Gene Symbol
- Psme2b
- Entrez Gene ID
- 621823
- Gene Name
- protease (prosome, macropain) activator subunit 2B
- Gene Aliases
- PA28b-p, PA28b2, Psme2-like, Psme2b-ps
- Chromosome Location
- Chr. 11: 48945345 - 48946410 on Build GRCm38
- UniGene ID
- Mm.440151
- Species
- Mus musculus
View Full Product Details