Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290122.1 | NP_001277051.1 | 3 | 763 | |
NM_001290123.1 | NP_001277052.1 | 3 | 752 | ||
NM_009032.3 | NP_033058.2 | 3 | 799 | ||
GenBank mRNA | AK049659.1 | - | 3 | 760 | |
AK194488.1 | - | 1 | 133 | ||
AK209057.1 | - | 1 | 133 | ||
BC130256.1 | - | 3 | 639 | 40142593 | |
BC144949.1 | - | 3 | 603 | 9053497 | |
U89506.1 | - | 4 | 627 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290127.1 | NP_001277056.1 | 3 | 1032 | |
NM_001290128.1 | NP_001277057.1 | 2 | 609 | ||
GenBank mRNA | EU287940.1 | - | 2 | 483 | |
EU287941.1 | - | 3 | 906 |