Gene Transcripts

Gene Symbol
Baiap2l2
Entrez Gene ID
207495
Gene Name
BAI1-associated protein 2-like 2
Gene Aliases
BC048937
Chromosome Location
Chr. 15: 79258195 - 79285532 on Build GRCm38
UniGene ID
Mm.309647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177580.3 NP_808248.1 6 539
GenBank mRNA AK143783.1 - 6 524
BC048937.1 - 6 477 6396012

View Full Product Details