Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_174993.2 | NP_778158.2 | 3 | 498 | |
GenBank mRNA | AY214130.1 | - | 3 | 403 | |
BC145139.1 | - | 3 | 479 | 9053689 | |
BC145140.1 | - | 3 | 479 | 9053690 | |
BC171947.1 | - | 3 | 479 | 9054233 | |
BC171948.1 | - | 3 | 479 | 9054234 |