Gene Transcripts

Gene Symbol
Stag2
Entrez Gene ID
20843
Gene Name
stromal antigen 2
Gene Aliases
9230105L23Rik, B230112I07Rik, SA-2, SAP2
Chromosome Location
Chr. X: 42149322 - 42277186 on Build GRCm38
UniGene ID
Mm.290422
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077712.2 NP_001071180.1 31 3913
NM_001290713.1 NP_001277642.1 31 3750
NM_021465.4 NP_067440.3 31 3773
GenBank mRNA AJ002636.1 - 30 3615
AK034759.1 - 8 1105
AK161072.1 - 3 379
BC066041.1 - 31 3654
CN708963.1 - 3 384

View Full Product Details