Gene Transcripts

Gene Symbol
Klhl32
Entrez Gene ID
212390
Gene Name
kelch-like 32
Gene Aliases
6430524H05Rik, D4Ertd389e, Gm1356, mKIAA1900
Chromosome Location
Chr. 4: 24614352 - 24851110 on Build GRCm38
UniGene ID
Mm.412074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033531.3 NP_001028703.2 4, 5 728
GenBank mRNA AK134944.1 - 4, 5 700

View Full Product Details