Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040400.2 | NP_001035490.2 | 3551 | ||
GenBank mRNA | AK151359.1 | - | 1, 2 | 541 | |
AK152297.1 | - | 1, 2 | 541 | ||
AK153251.1 | - | 1, 2 | 541 | ||
AK153311.1 | - | 1, 2 | 541 | ||
AK153460.1 | - | 1, 2 | 541 | ||
DQ079067.1 | - | 3, 4 | 3529 | ||
GU079949.1 | - | 1, 2 | 3175 |