Gene Transcripts

Gene Symbol
Prss40
Entrez Gene ID
21756
Gene Name
protease, serine 40
Gene Aliases
Tesp2
Chromosome Location
Chr. 1: 34536515 - 34560993 on Build GRCm38
UniGene ID
Mm.14418
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009356.2 NP_033382.2 1 187
GenBank mRNA AB008911.1 - 1 170
AK132909.1 - 1 188
BC145989.1 - 1 202 40131292

View Full Product Details