Gene Transcripts

Gene Symbol
Speer2
Entrez Gene ID
224318
Gene Name
spermatogenesis associated glutamate (E)-rich protein 2
Gene Aliases
SPEER-2
Chromosome Location
Chr. 16: 69856874 - 69863794 on Build GRCm38
UniGene ID
Mm.212743
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173069.3 NP_775092.3 3, 4 444
GenBank mRNA AY151401.1 - 2, 3 215
BB610282.1 - 3, 4 446

View Full Product Details