Gene Transcripts

Gene Symbol
Obp2a
Entrez Gene ID
227627
Gene Name
odorant binding protein 2A
Gene Aliases
BC027556, Lcn13
Chromosome Location
Chr. 2: 25697543 - 25703326 on Build GRCm38
UniGene ID
Mm.44630
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153558.1 NP_705786.1 2 162
GenBank mRNA AY360148.1 - 2 198
BC027556.1 - 2 162 1434737

View Full Product Details