Gene Transcripts

Gene Symbol
Cyb5rl
Entrez Gene ID
230582
Gene Name
cytochrome b5 reductase-like
Gene Aliases
2810410C14Rik, mFLJ00377
Chromosome Location
Chr. 4: 107066844 - 107087993 on Build GRCm38
UniGene ID
Mm.44608
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175471.2 NP_780680.1 4 694
GenBank mRNA AK042316.1 - 5 2309
AK220154.1 - 5 596
BC043687.1 - 4 694 5136046

View Full Product Details