Gene Transcripts

Gene Symbol
Glis1
Entrez Gene ID
230587
Gene Name
GLIS family zinc finger 1
Gene Aliases
Gli5, Gli6, GliH1
Chromosome Location
Chr. 4: 107426640 - 107635061 on Build GRCm38
UniGene ID
Mm.331757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147221.2 NP_671754.2 4 848
GenBank mRNA AF220434.1 - 2 702
BC066157.1 - 2 283 30468539
JQ043365.1 - 3 627

View Full Product Details