Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198027.2 | NP_932144.2 | 2, 3 | 169 | |
GenBank mRNA | AK082058.1 | - | 2, 3 | 135 | |
BC029805.1 | - | 2, 3 | 121 | 4952089 | |
BC080310.1 | - | 1, 2 | 90 | ||
BY196893.1 | - | 2, 3 | 170 |